Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LEMD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167614.1 | NP_001161086.1 | 1 | 577 | ||
NM_014319.4 | NP_055134.2 | 1 | 577 | |||
GenBank mRNA | AF112299.2 | - | 1 | 557 | ||
AK304317.1 | - | 1 | 565 | |||
AK312473.1 | - | 1 | 577 |