Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014329.4 | NP_055144.3 | 2 | 413 | ||
GenBank mRNA | AK291049.1 | - | 2 | 181 | ||
AK302563.1 | - | 2 | 179 | |||
BC043616.1 | - | 2 | 316 | 6065418 | ||
BC064567.1 | - | 2 | 310 | 5575195 | ||
BX445706.2 | - | 2 | 365 | |||
DC403765.1 | - | 2 | 414 | |||
L26339.1 | - | 2 | 310 |