Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CLEC4M SIRNA Assays ›
Gene Symbol:
CLEC4M
Gene Name:
C-type lectin domain family 4 member M
Gene Aliases:
CD209L, CD299, DC-SIGN2, DC-SIGNR, DCSIGNR, HP10347, L-SIGN, LSIGN
Chromosome Location:
Chr.19: 7763052 - 7770559 on Build GRCh38
UniGene ID:
Hs.421437
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
21073
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CLEC4M
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001144904.1
NP_001138376.1
5
783
NM_001144905.1
NP_001138377.1
6
864
NM_001144906.1
NP_001138378.1
4
528
NM_001144907.1
NP_001138379.1
5
735
NM_001144908.1
NP_001138380.1
5
660
NM_001144909.1
NP_001138381.1
6
798
NM_001144910.1
NP_001138382.1
6
867
NM_001144911.1
NP_001138383.1
4
852
NM_014257.4
NP_055072.3
5
936
NR_026707.1
4
1384
NR_026708.1
4
1384
NR_026709.1
3
1321
XM_006722611.2
XP_006722674.1
5
868
XM_006722612.3
XP_006722675.1
4
942
XM_006722613.3
XP_006722676.1
4
944
XM_006722614.3
XP_006722677.1
3
860
XM_006722615.2
XP_006722678.1
5
936
GenBank mRNA
AB015629.1
-
4
758
AF245219.2
-
5
842
AF290887.1
-
6
802
AK023750.1
-
2
1105
AK292278.1
-
6
863
AK304499.1
-
5
716
AK314704.1
-
6
797
AY042234.1
-
5
818
AY042235.1
-
4
1266
AY042236.1
-
4
1266
AY042237.1
-
5
542
AY042238.1
-
5
617
AY042239.1
-
3
1203
AY042240.1
-
4
410
AY343913.1
-
6
681
BC038851.1
-
5
890
5166966
BC110614.2
-
3
791
40032453
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane traffic protein
Gene Ontology Categories:
Function(s)
Process(es)
adaptive immune response
endocytosis
leukocyte cell-cell adhesion
cell-cell recognition
regulation of gene expression
modulation by virus of host morphology or physiology
virion attachment to host cell
viral genome replication
antigen processing and presentation
regulation of blood coagulation
intracellular signal transduction
innate immune response
viral entry into host cell
peptide antigen transport
intracellular transport of virus
virus receptor activity
receptor activity
mannose binding
carbohydrate binding
ICAM-3 receptor activity
peptide antigen binding
virion binding
metal ion binding
calcium-dependent protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0