Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PFDN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185181.2 | NP_001172110.1 | 3 | 466 | ||
NM_001265595.1 | NP_001252524.1 | 4 | 358 | |||
NM_001265596.1 | NP_001252525.1 | 4 | 355 | |||
NM_014260.3 | NP_055075.1 | 4 | 363 | |||
GenBank mRNA | AK312150.1 | - | 4 | 351 | ||
BC039033.1 | - | 3 | 332 | 5748799 | ||
BC059783.1 | - | 3 | 338 | 5415221 | ||
BE794658.1 | - | 3 | 319 | |||
BM686351.1 | - | 3 | 447 | |||
BM745352.1 | - | 4 | 358 | |||
BM792413.1 | - | 3 | 340 | |||
BM818422.1 | - | 4 | 358 | |||
BM834357.1 | - | 4 | 359 | |||
DA256816.1 | - | 4 | 351 |