Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PES1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243225.1 | NP_001230154.1 | 4 | 397 | ||
NM_001282327.1 | NP_001269256.1 | 6 | 795 | |||
NM_001282328.1 | NP_001269257.1 | 6 | 842 | |||
NM_014303.3 | NP_055118.1 | 4 | 397 | |||
XM_006724173.1 | XP_006724236.1 | 4 | 353 | |||
GenBank mRNA | AB451380.1 | - | 4 | 290 | ||
AK096320.1 | - | 6 | 814 | |||
AK127185.1 | - | 6 | 796 | |||
AK315518.1 | - | 4 | 364 | |||
AU130886.1 | - | 4 | 364 | |||
BC032489.1 | - | 4 | 354 | 5416478 | ||
CR456539.1 | - | 4 | 315 | |||
U78310.1 | - | 4 | 348 |