Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014314.3 | NP_055129.2 | 2 | 277 | ||
GenBank mRNA | AF038963.1 | - | 2 | 276 | ||
AK125989.1 | - | 2 | 227 | |||
AK301678.1 | - | 2 | 148 | |||
AK315040.1 | - | 2 | 148 | |||
BC107731.1 | - | 2 | 299 | |||
BC132786.1 | - | 2 | 225 | 40146808 | ||
BC136610.1 | - | 2 | 225 | 9020600 | ||
DN998145.1 | - | 2 | 278 |