Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001326380.1 | NP_001313309.1 | 3 | 366 | ||
NM_014320.2 | NP_055135.1 | 3 | 597 | |||
GenBank mRNA | AF117616.1 | - | 3 | 575 | ||
AF411610.1 | - | 3 | 237 | |||
AK057928.1 | - | 3 | 512 | |||
AY427823.1 | - | 3 | 479 | |||
BC008205.1 | - | 3 | 461 | 3869026 | ||
BC010290.1 | - | 3 | 474 | 3445763 | ||
BC016806.1 | - | 3 | 577 | |||
BC016807.1 | - | 3 | 577 | |||
BC093037.1 | - | 3 | 460 | 30333215 |