Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RUSC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105203.1 | NP_001098673.1 | 10 | 3119 | ||
NM_001105204.1 | NP_001098674.1 | 10 | 2801 | |||
NM_001105205.1 | NP_001098675.1 | 9 | 2141 | |||
NM_001278227.1 | NP_001265156.1 | 9 | 2085 | |||
NM_001278228.1 | NP_001265157.1 | 8 | 2321 | |||
NM_001278229.1 | NP_001265158.1 | 9 | 1866 | |||
NM_001278230.1 | NP_001265159.1 | 8 | 1983 | |||
NM_014328.4 | NP_055143.2 | 9 | 2184 | |||
NR_103478.1 | 8 | 1531 | ||||
XM_006711254.1 | XP_006711317.1 | 11 | 3530 | |||
XM_006711256.1 | XP_006711319.1 | 11 | 3212 | |||
XM_006711257.1 | XP_006711320.1 | 9 | 1798 | |||
XM_017000891.1 | XP_016856380.1 | 10 | 3700 | |||
XM_017000892.1 | XP_016856381.1 | 9 | 1480 | |||
GenBank mRNA | AB026894.2 | - | 9 | 1608 | ||
AK074485.1 | - | 8 | 1419 | |||
AK074982.1 | - | 9 | 1603 | |||
AK125378.1 | - | 10 | 2753 | |||
AK298730.1 | - | 9 | 1595 | |||
AL080083.1 | - | 8 | 1461 | |||
BC001045.2 | - | 9 | 1625 | |||
BC004293.1 | - | 8 | 1387 | |||
BX640612.1 | - | 8 | 1801 | |||
CR749671.1 | - | 8 | 2690 |