Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LPAR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162497.1 | NP_001155969.1 | 5 | 1631 | ||
NM_001162498.1 | NP_001155970.1 | 1 | 1582 | |||
NM_005767.5 | NP_005758.2 | 7 | 2113 | |||
GenBank mRNA | AF000546.1 | - | 1 | 532 | ||
AK122856.1 | - | 5 | 1631 | |||
AK123059.1 | - | 1 | 1178 | |||
AK223389.1 | - | 1 | 947 | |||
BC040850.1 | - | 1 | 1165 | 5180625 | ||
BC045651.1 | - | 7 | 2114 | 5269149 | ||
BC070295.1 | - | 1 | 994 | 6663864 | ||
BC106756.2 | - | 1 | 697 | 40024517 | ||
BX537392.1 | - | 1 | 1178 | |||
BX537438.1 | - | 1 | 1178 | |||
BX537445.1 | - | 1 | 1178 | |||
BX647109.1 | - | 1 | 1178 | |||
HQ995530.1 | - | 1 | 488 | |||
JF810890.1 | - | 1 | 488 |