Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014329.4 | NP_055144.3 | 7 | 1111 | ||
GenBank mRNA | AK128582.1 | - | 7 | 1265 | ||
AK291049.1 | - | 7 | 879 | |||
AK302563.1 | - | 7 | 877 | |||
BC043616.1 | - | 7 | 1014 | 6065418 | ||
BC053598.1 | - | 2 | 251 | |||
BC064567.1 | - | 7 | 1008 | 5575195 | ||
L26339.1 | - | 7 | 1008 |