Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CADM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098517.1 | NP_001091987.1 | 1, 2 | 261 | ||
NM_001301043.1 | NP_001287972.1 | 1, 2 | 261 | |||
NM_001301044.1 | NP_001287973.1 | 1, 2 | 261 | |||
NM_001301045.1 | NP_001287974.1 | 1, 2 | 261 | |||
NM_014333.3 | NP_055148.3 | 1, 2 | 261 | |||
XM_005271494.2 | XP_005271551.1 | 1, 2 | 154 | |||
GenBank mRNA | AB094146.1 | - | 1, 2 | 132 | ||
AF132811.1 | - | 1, 2 | 135 | |||
AK075502.1 | - | 1, 2 | 151 | |||
AK315245.1 | - | 1, 2 | 194 | |||
AY358334.1 | - | 1, 2 | 147 | |||
BC035930.1 | - | 1, 2 | 153 | |||
BC047021.1 | - | 1, 2 | 261 | 5248100 | ||
BC125102.1 | - | 1, 2 | 136 | |||
BC125103.1 | - | 1, 2 | 135 | 40118330 | ||
BI915307.1 | - | 1, 2 | 261 | |||
CR457157.1 | - | 1, 2 | 132 | |||
HE586496.1 | - | 1, 2 | 135 | |||
HE586497.1 | - | 1, 2 | 135 | |||
HE586498.1 | - | 1, 2 | 135 | |||
HE586499.1 | - | 1, 2 | 135 | |||
KJ534791.1 | - | 1, 2 | 132 | |||
KJ534792.1 | - | 1, 2 | 132 | |||
KJ534793.1 | - | 1, 2 | 132 | |||
KJ534794.1 | - | 1, 2 | 132 | |||
KJ534795.1 | - | 1, 2 | 132 | |||
KJ534796.1 | - | 1, 2 | 132 | |||
KJ534797.1 | - | 1, 2 | 132 |