Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GHITM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014394.2 | NP_055209.2 | 2, 3 | 340 | ||
GenBank mRNA | AB009685.1 | - | 1, 2 | 136 | ||
AF060923.1 | - | 2, 3 | 214 | |||
AF078863.1 | - | 2, 3 | 265 | |||
AF131820.1 | - | 2, 3 | 267 | |||
AF161400.1 | - | 2, 3 | 249 | |||
AK098770.1 | - | 2, 3 | 213 | |||
AK130094.1 | - | 2, 3 | 243 | |||
AK130097.1 | - | 2, 3 | 208 | |||
AK225318.1 | - | 2, 3 | 217 | |||
AK292864.1 | - | 2, 3 | 208 | |||
AK297960.1 | - | 2, 3 | 207 | |||
AK299937.1 | - | 246 | ||||
AL136713.1 | - | 2, 3 | 250 | |||
AY358824.1 | - | 2, 3 | 215 | |||
BC010354.1 | - | 2, 3 | 211 | 4093037 | ||
BC022872.1 | - | 2, 3 | 222 | |||
CR533519.1 | - | 1, 2 | 136 |