Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DKK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014421.2 | NP_055236.1 | 2 | 959 | ||
GenBank mRNA | AB033208.1 | - | 2 | 476 | ||
AB033941.1 | - | 2 | 476 | |||
AF177395.1 | - | 2 | 971 | |||
AK309466.1 | - | 3 | 188 | |||
AK312695.1 | - | 2 | 485 | |||
AY358414.1 | - | 2 | 464 | |||
BC075077.2 | - | 2 | 288 | 30915382 | ||
BC075078.2 | - | 2 | 288 | 30915595 | ||
BC126328.1 | - | 2 | 560 | 8992044 | ||
BC126330.1 | - | 2 | 560 | 8992046 |