Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIR3DS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083539.2 | NP_001077008.1 | 3 | 234 | ||
GenBank mRNA | AF022044.1 | - | 3 | 210 | ||
AJ417558.3 | - | 1 | 146 | |||
AK301140.1 | - | 3 | 234 | |||
BC098288.1 | - | 3 | 208 | 40026980 | ||
BC105679.1 | - | 3 | 208 | 40004313 | ||
DC390157.1 | - | 3 | 234 | |||
EF370406.1 | - | 2 | 158 | |||
EF582383.1 | - | 3 | 197 | |||
L76661.1 | - | 3 | 208 | |||
U73396.1 | - | 3 | 197 | |||
X97233.1 | - | 3 | 197 |
Gene Symbol: KIR3DL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322168.1 | NP_001309097.1 | 3 | 260 | ||
GenBank mRNA | AF022049.1 | - | 3 | 197 | ||
AF103010.1 | - | 3 | 197 | |||
AF103011.1 | - | 3 | 197 | |||
AF103012.1 | - | 3 | 197 | |||
AF103013.1 | - | 3 | 197 | |||
AF208688.1 | - | 1 | 134 | |||
AF208689.1 | - | 1 | 134 | |||
AF262972.1 | - | 1 | 116 | |||
AF262973.1 | - | 3 | 230 | |||
AF262974.1 | - | 3 | 230 | |||
AJ417556.1 | - | 1 | 146 | |||
AJ417557.1 | - | 1 | 146 | |||
AY366236.1 | - | 3 | 235 | |||
AY366249.1 | - | 3 | 197 | |||
AY366251.1 | - | 3 | 197 | |||
AY366252.1 | - | 3 | 197 | |||
AY760050.1 | - | 3 | 197 | |||
AY789065.1 | - | 3 | 212 | |||
AY789066.1 | - | 3 | 197 | |||
BC028206.1 | - | 3 | 224 | 5215674 | ||
BC101540.1 | - | 3 | 216 | 8069046 | ||
BC101542.1 | - | 3 | 216 | 8069048 | ||
EF462885.1 | - | 1 | 127 | |||
EF472674.1 | - | 1 | 127 | |||
EF472675.1 | - | 1 | 127 | |||
EF472676.1 | - | 1 | 127 | |||
EF472678.1 | - | 1 | 127 | |||
EF472679.1 | - | 1 | 127 | |||
U30273.1 | - | 3 | 199 | |||
U31416.1 | - | 3 | 222 | |||
U33328.1 | - | 3 | 222 |