Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282454.1 | NP_001269383.1 | 3 | 449 | ||
NM_014426.3 | NP_055241.1 | 3 | 577 | |||
NM_152227.2 | NP_689413.1 | 4 | 532 | |||
GenBank mRNA | AF121855.1 | - | 4 | 388 | ||
AK001793.1 | - | 3 | 294 | |||
AK054634.1 | - | 3 | 1013 | |||
AK129505.1 | - | 3 | 294 | |||
AK223308.1 | - | 3 | 262 | |||
AK296947.1 | - | 4 | 401 | |||
AK308017.1 | - | 4 | 533 | |||
BC062638.1 | - | 3 | 234 | |||
BC093623.1 | - | 3 | 503 | 7939468 | ||
BC093980.1 | - | 3 | 503 | 7939825 | ||
BC143274.1 | - | 3 | 503 | 9051776 | ||
DA567854.1 | - | 4 | 532 |