Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VDAC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184783.2 | NP_001171712.1 | 7 | 870 | ||
NM_001184823.1 | NP_001171752.1 | 5 | 295 | |||
NM_001324087.1 | NP_001311016.1 | 7 | 796 | |||
NM_001324088.1 | NP_001311017.1 | 6 | 470 | |||
NM_001324089.1 | NP_001311018.1 | 7 | 574 | |||
NM_001324090.1 | NP_001311019.1 | 7 | 738 | |||
NM_003375.4 | NP_003366.2 | 6 | 692 | |||
GenBank mRNA | AK296635.1 | - | 6 | 386 | ||
AK315919.1 | - | 6 | 450 | |||
BC000165.2 | - | 5 | 370 | 2901130 | ||
BC012883.1 | - | 5 | 328 | 3875077 | ||
BC072407.1 | - | 5 | 360 | 6137879 | ||
BE545584.1 | - | 5 | 292 | |||
BU171142.1 | - | 1 | 278 | |||
CR456964.1 | - | 3 | 178 | |||
L06328.1 | - | 4 | 273 | |||
L08666.1 | - | 6 | 503 |