Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPHA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005232.4 | NP_005223.4 | 13 | 2183 | ||
XM_006715880.3 | XP_006715943.1 | 12 | 1811 | |||
GenBank mRNA | AK313263.1 | - | 13 | 2156 | ||
BC130291.1 | - | 13 | 2167 | 40146322 | ||
BC144156.1 | - | 12 | 2319 | |||
CN289901.1 | - | 6 | 544 | |||
M18391.1 | - | 13 | 2213 | |||
Z27409.1 | - | 9 | 1241 |