Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144932.2 | NP_001138404.1 | 1 | 425 | ||
NM_002797.4 | NP_002788.1 | 1 | 425 | |||
XM_005267871.2 | XP_005267928.1 | 1 | 420 | |||
GenBank mRNA | AK225888.1 | - | 1 | 190 | ||
AK300714.1 | - | 1 | 170 | |||
AK311895.1 | - | 1 | 181 | |||
BC057840.1 | - | 1 | 180 | 4795732 | ||
BC107720.1 | - | 1 | 176 | 6738409 | ||
BX248299.1 | - | 1 | 157 | |||
BX538001.1 | - | 1 | 172 | |||
S74378.1 | - | 1 | 17 |