Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP2R5D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270476.1 | NP_001257405.1 | 4, 5 | 714 | ||
NM_006245.3 | NP_006236.1 | 4, 5 | 697 | |||
NM_180976.2 | NP_851307.1 | 4, 5 | 601 | |||
NM_180977.2 | NP_851308.1 | 2, 3 | 379 | |||
GenBank mRNA | AB000634.1 | - | 4, 5 | 532 | ||
AB000635.1 | - | 2, 3 | 226 | |||
AB209861.1 | - | 2, 3 | 276 | |||
AB451342.1 | - | 4, 5 | 518 | |||
AB451357.1 | - | 4, 5 | 518 | |||
AK122701.1 | - | 4, 5 | 591 | |||
AK290604.1 | - | 4, 5 | 510 | |||
AK291797.1 | - | 4, 5 | 605 | |||
AK299696.1 | - | 4, 5 | 570 | |||
BC001095.2 | - | 4, 5 | 581 | 3507280 | ||
BC001175.1 | - | 4, 5 | 508 | 3355395 | ||
BC010692.1 | - | 4, 5 | 591 | 3853892 | ||
D78360.1 | - | 4, 5 | 499 | |||
L76702.1 | - | 4, 5 | 705 |