Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4GALT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005417.2 | NP_001005417.1 | 7 | 1227 | ||
NM_003780.4 | NP_003771.1 | 7 | 1427 | |||
NM_030587.2 | NP_085076.2 | 7 | 1228 | |||
XM_017002716.1 | XP_016858205.1 | 7 | 1185 | |||
XM_017002717.1 | XP_016858206.1 | 7 | 1388 | |||
GenBank mRNA | AB024434.1 | - | 6 | 1073 | ||
AF038660.1 | - | 6 | 1117 | |||
AK074533.1 | - | 5 | 725 | |||
AK095873.1 | - | 7 | 1855 | |||
AK293418.1 | - | 7 | 1177 | |||
AL137647.1 | - | 5 | 622 | |||
BC096821.1 | - | 7 | 1195 | 30558762 | ||
Y12510.1 | - | 6 | 1065 |