Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPN10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023083.3 | NP_075571.1 | 1, 2 | 345 | ||
NM_023085.3 | NP_075573.2 | 1, 2 | 345 | |||
GenBank mRNA | AB058748.1 | - | 1, 2 | 249 | ||
AF089088.1 | - | 1, 2 | 326 | |||
AF089090.1 | - | 1, 2 | 190 | |||
AF089091.1 | - | 1, 2 | 190 | |||
AF089092.1 | - | 1, 2 | 190 | |||
AF089093.1 | - | 1, 2 | 190 | |||
AF089094.1 | - | 1, 2 | 190 | |||
AK074807.1 | - | 1, 2 | 296 | |||
AK300279.1 | - | 1, 2 | 327 | |||
AL558905.3 | - | 1, 2 | 343 | |||
BC004260.1 | - | 1, 2 | 294 | 3606959 | ||
BC007553.1 | - | 1, 2 | 294 | 2967328 | ||
DA413480.1 | - | 1, 2 | 336 | |||
DA623373.1 | - | 1, 2 | 333 |