Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143679.1 | NP_001137151.1 | 7 | 1134 | ||
NM_001143680.1 | NP_001137152.1 | 7 | 930 | |||
NM_001143681.1 | NP_001137153.1 | 7 | 837 | |||
NM_015917.2 | NP_057001.1 | 8 | 966 | |||
GenBank mRNA | AF068287.1 | - | 8 | 952 | ||
AF070657.1 | - | 8 | 924 | |||
AF087849.1 | - | 8 | 938 | |||
AK000640.1 | - | 8 | 924 | |||
AK289580.1 | - | 8 | 904 | |||
AK295592.1 | - | 7 | 825 | |||
AK299968.1 | - | 7 | 848 | |||
AL136938.1 | - | 8 | 952 | |||
AY520571.1 | - | 8 | 935 | |||
BC001231.1 | - | 8 | 913 | 3460150 | ||
BC050715.1 | - | 8 | 886 | 6061265 | ||
BC063425.1 | - | 7 | 1063 | 5218251 |