Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIR2DS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281971.1 | NP_001268900.1 | 8 | 1543 | ||
NM_001281972.1 | NP_001268901.1 | 7 | 1492 | |||
NM_012314.5 | NP_036446.3 | 9 | 1565 | |||
GenBank mRNA | U24077.1 | - | 8 | 1467 |
Gene Symbol: KIR2DS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291695.1 | NP_001278624.1 | 8 | 1445 | ||
NM_001291696.1 | NP_001278625.1 | 7 | 1211 | |||
NM_001291700.1 | NP_001278629.1 | 6 | 917 | |||
NM_001291701.1 | NP_001278630.1 | 7 | 1460 | |||
NM_012312.4 | NP_036444.1 | 8 | 1511 | |||
GenBank mRNA | L41347.1 | - | 8 | 1495 | ||
U24079.1 | - | 8 | 1474 |