Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNMT3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320892.1 | NP_001307821.1 | 3 | 368 | ||
NM_022552.4 | NP_072046.2 | 3 | 368 | |||
NM_175629.2 | NP_783328.1 | 3 | 439 | |||
NM_175630.1 | NP_783329.1 | 3 | 439 | |||
NR_135490.1 | 3 | 439 | ||||
XM_005264175.4 | XP_005264232.1 | 3 | 302 | |||
XM_011532663.2 | XP_011530965.1 | 3 | 378 | |||
XM_011532664.2 | XP_011530966.1 | 3 | 379 | |||
XM_017003526.1 | XP_016859015.1 | 3 | 410 | |||
GenBank mRNA | AB208833.1 | - | 3 | 338 | ||
AF067972.2 | - | 3 | 330 | |||
AF331856.1 | - | 3 | 318 | |||
AL543971.3 | - | 3 | 367 | |||
BC032392.1 | - | 3 | 439 | |||
BC043617.1 | - | 3 | 338 | 6150112 | ||
BC051864.1 | - | 3 | 307 |