Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002536.2 | NP_002527.1 | 3 | 401 | ||
GenBank mRNA | AB449912.1 | - | 3 | 290 | ||
AK293521.1 | - | 3 | 359 | |||
AK293875.1 | - | 3 | 360 | |||
AK294776.1 | - | 3 | 299 | |||
AK298884.1 | - | 3 | 388 | |||
BC101817.1 | - | 3 | 327 | 8069323 | ||
BC101819.1 | - | 3 | 327 | 8069325 | ||
BC125088.1 | - | 3 | 316 | 40117866 | ||
DA087200.1 | - | 3 | 401 | |||
L08240.1 | - | 2 | 180 |