Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPT1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004377.3 | NP_004368.1 | 19 | 2616 | ||
NM_152245.2 | NP_689451.1 | 19 | 2655 | |||
GenBank mRNA | U66828.1 | - | 19 | 2555 | ||
Y08683.1 | - | 19 | 2568 |