Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIR3DL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322168.1 | NP_001309097.1 | 5 | 780 | ||
NM_013289.2 | NP_037421.2 | 5 | 780 | |||
GenBank mRNA | AF022049.1 | - | 5 | 717 | ||
AF103010.1 | - | 5 | 717 | |||
AF103011.1 | - | 5 | 717 | |||
AF103012.1 | - | 5 | 717 | |||
AF103013.1 | - | 5 | 717 | |||
AF208689.1 | - | 3 | 654 | |||
AF262968.1 | - | 3 | 636 | |||
AF262969.1 | - | 3 | 636 | |||
AF262970.1 | - | 3 | 636 | |||
AF262971.1 | - | 3 | 636 | |||
AF262972.1 | - | 3 | 636 | |||
AF262973.1 | - | 5 | 750 | |||
AF262974.1 | - | 5 | 750 | |||
AJ417556.1 | - | 3 | 666 | |||
AJ417557.1 | - | 3 | 666 | |||
AY366236.1 | - | 5 | 755 | |||
AY366249.1 | - | 5 | 717 | |||
AY366250.1 | - | 5 | 743 | |||
AY366251.1 | - | 5 | 717 | |||
AY366252.1 | - | 5 | 717 | |||
AY760050.1 | - | 5 | 717 | |||
AY789065.1 | - | 5 | 732 | |||
AY789066.1 | - | 5 | 717 | |||
AY789072.1 | - | 5 | 717 | |||
BC028206.1 | - | 5 | 744 | 5215674 | ||
BC101540.1 | - | 5 | 736 | 8069046 | ||
BC101542.1 | - | 5 | 736 | 8069048 | ||
DQ178659.1 | - | 5 | 717 | |||
EF462885.1 | - | 3 | 647 | |||
EF472674.1 | - | 3 | 647 | |||
EF472675.1 | - | 3 | 647 | |||
EF472676.1 | - | 3 | 647 | |||
EF472677.1 | - | 3 | 647 | |||
EF472678.1 | - | 3 | 647 | |||
EF472679.1 | - | 3 | 647 | |||
L41269.1 | - | 5 | 736 | |||
L76664.1 | - | 4 | 443 | |||
U30273.1 | - | 5 | 719 | |||
U30274.1 | - | 5 | 780 | |||
U31416.1 | - | 5 | 742 | |||
U33328.1 | - | 5 | 742 | |||
X94262.1 | - | 5 | 718 |
Gene Symbol: 1060P11.3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ630586.1 | - | 5 | 718 |
Gene Symbol: KIR2DL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014219.2 | NP_055034.2 | 4 | 461 | ||
GenBank mRNA | AF285434.1 | - | 3 | 392 | ||
AY366242.1 | - | 4 | 461 | |||
KP260924.1 | - | 4 | 428 | |||
L76669.1 | - | 4 | 443 | |||
U24075.1 | - | 4 | 446 |
Gene Symbol: KIR2DL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015868.2 | NP_056952.2 | 4 | 469 | ||
GenBank mRNA | AF022048.1 | - | 4 | 432 | ||
AF285435.1 | - | 3 | 392 | |||
AY789057.1 | - | 4 | 432 | |||
BC032422.1 | - | 4 | 445 | 5220339 | ||
BC050730.1 | - | 4 | 463 | 5219336 | ||
BC110613.1 | - | 4 | 446 | 40031619 | ||
FN395076.1 | - | 4 | 432 | |||
FN552550.1 | - | 4 | 414 | |||
L41268.1 | - | 4 | 469 | |||
L76662.1 | - | 4 | 443 | |||
L76663.1 | - | 4 | 443 | |||
U24074.1 | - | 4 | 453 | |||
U73395.1 | - | 4 | 453 | |||
U96190.1 | - | 2 | 369 |
Gene Symbol: KIR2DS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291695.1 | NP_001278624.1 | 4 | 467 | ||
NM_001291696.1 | NP_001278625.1 | 3 | 167 | |||
NM_001291701.1 | NP_001278630.1 | 4 | 467 | |||
NM_012312.4 | NP_036444.1 | 4 | 467 | |||
GenBank mRNA | AF285438.1 | - | 3 | 392 | ||
AJ002102.1 | - | 4 | 429 | |||
AJ002103.1 | - | 4 | 432 | |||
AJ002104.1 | - | 4 | 432 | |||
AJ002105.1 | - | 4 | 432 | |||
AM409094.1 | - | 4 | 433 | |||
AY366243.1 | - | 4 | 442 | |||
AY366244.1 | - | 4 | 431 | |||
BC108917.1 | - | 4 | 434 | 40017299 | ||
BC119631.1 | - | 4 | 451 | 40115568 | ||
BC119632.1 | - | 4 | 451 | 40115570 | ||
BC125146.1 | - | 3 | 416 | |||
BC126856.1 | - | 4 | 434 | 40017301 | ||
FR750994.1 | - | 4 | 467 | |||
L41347.1 | - | 4 | 451 | |||
L76668.1 | - | 3 | 143 | |||
U24079.1 | - | 4 | 430 | |||
X89893.1 | - | 4 | 432 |
Gene Symbol: KIR2DS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012313.1 | NP_036445.1 | 4 | 443 | ||
GenBank mRNA | AF022047.1 | - | 4 | 436 | ||
AM851002.1 | - | 4 | 468 | |||
AY789063.2 | - | 3 | 416 | |||
L76670.1 | - | 4 | 443 | |||
X97231.1 | - | 4 | 491 |
Gene Symbol: KIR2DS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014513.2 | NP_055328.2 | 4 | 466 | ||
GenBank mRNA | AF208054.1 | - | 4 | 466 | ||
AY366247.1 | - | 4 | 490 | |||
AY366248.1 | - | 5 | 490 | |||
L76672.1 | - | 4 | 443 |
Gene Symbol: KIR3DS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083539.2 | NP_001077008.1 | 5 | 754 | ||
NM_001282170.1 | NP_001269099.1 | 4 | 469 | |||
NM_001282171.1 | NP_001269100.1 | 3 | 433 | |||
GenBank mRNA | AF022044.1 | - | 5 | 730 | ||
AJ417558.3 | - | 3 | 666 | |||
AK301140.1 | - | 5 | 754 | |||
BC098107.1 | - | 3 | 407 | 40026977 | ||
BC098288.1 | - | 5 | 728 | 40026980 | ||
BC105679.1 | - | 5 | 728 | 40004313 | ||
EF370406.1 | - | 4 | 677 | |||
EF582383.1 | - | 5 | 717 | |||
L76661.1 | - | 5 | 728 | |||
U73396.1 | - | 5 | 717 | |||
X97232.1 | - | 4 | 432 | |||
X97233.1 | - | 5 | 717 |