Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000075.3 | NP_000066.1 | 8 | 1146 | ||
GenBank mRNA | AK297901.1 | - | 7 | 889 | ||
AK313701.1 | - | 8 | 970 | |||
BC003644.2 | - | 8 | 993 | 3532812 | ||
BC005864.2 | - | 8 | 1057 | 2961345 | ||
BC010153.2 | - | 8 | 995 | 3531300 | ||
BC015669.2 | - | 7 | 679 | |||
CR407668.1 | - | 7 | 854 | |||
CR542247.1 | - | 7 | 854 | |||
M14505.1 | - | 8 | 1081 | |||
U79269.1 | - | 8 | 941 | |||
Z48970.1 | - | 8 | 988 |