Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014987.1 | NP_001014987.1 | 7 | 726 | ||
NM_001014988.1 | NP_001014988.1 | 7 | 723 | |||
NM_001014989.1 | NP_001014989.2 | 8 | 518 | |||
NM_014387.3 | NP_055202.1 | 6 | 813 | |||
GenBank mRNA | AF036905.1 | - | 7 | 431 | ||
AF036906.1 | - | 6 | 539 | |||
AJ223280.1 | - | 7 | 696 | |||
BC011563.1 | - | 8 | 518 | 4644259 | ||
BI911769.1 | - | 7 | 692 | |||
FN432831.1 | - | 6 | 488 | |||
FN432832.1 | - | 6 | 458 | |||
FN432833.1 | - | 5 | 431 | |||
FN432834.1 | - | 6 | 345 |