Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRDM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198.3 | NP_001189.2 | 7 | 4834 | ||
NM_182907.2 | NP_878911.1 | 5 | 4422 | |||
XM_006715550.3 | XP_006715613.1 | 6 | 5683 | |||
XM_011536062.2 | XP_011534364.1 | 7 | 5808 | |||
XM_011536063.2 | XP_011534365.1 | 7 | 4886 | |||
XM_011536064.2 | XP_011534366.1 | 5 | 5987 | |||
XM_017011187.1 | XP_016866676.1 | 7 | 4621 | |||
GenBank mRNA | AK129768.1 | - | 1 | 1610 | ||
AY198414.1 | - | 7 | 4834 | |||
AY198415.1 | - | 5 | 4344 |