Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EZH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321079.1 | NP_001308008.1 | 3, 4 | 336 | ||
NM_001991.4 | NP_001982.2 | 4, 5 | 427 | |||
XM_017024352.1 | XP_016879841.1 | 4, 5 | 371 | |||
GenBank mRNA | AB002386.1 | - | 3, 4 | 339 | ||
AB004818.1 | - | 3, 4 | 345 | |||
AK295626.1 | - | 3, 4 | 289 | |||
AK304835.1 | - | 3, 4 | 240 | |||
AK309121.1 | - | 4, 5 | 368 | |||
BC015882.1 | - | 4, 5 | 361 | 3908776 | ||
BT009782.1 | - | 2, 3 | 239 | |||
DC333017.1 | - | 4, 5 | 384 | |||
DC423942.1 | - | 4, 5 | 427 | |||
U50315.1 | - | 3, 4 | 345 |