Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUV39H1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282166.1 | NP_001269095.1 | 6 | 2693 | ||
NM_003173.3 | NP_003164.1 | 6 | 2446 | |||
GenBank mRNA | AF019968.1 | - | 6 | 2438 | ||
BC006238.2 | - | 6 | 2437 | 3945632 | ||
L08238.1 | - | 5 | 2535 |