Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2G1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003342.4 | NP_003333.1 | 6 | 1588 | ||
GenBank mRNA | BC026288.1 | - | 6 | 1396 |
Gene Symbol: LOC103021295 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110982.2 | 1 | 58 | |||
GenBank mRNA | AW499792.1 | - | 1 | 64 | ||
DB524022.1 | - | 1 | 58 |