Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G3BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005754.2 | NP_005745.1 | 12 | 1539 | ||
NM_198395.1 | NP_938405.1 | 12 | 1508 | |||
XM_006714749.3 | XP_006714812.1 | 12 | 1528 | |||
XM_006714750.3 | XP_006714813.1 | 12 | 1467 | |||
XM_017008922.1 | XP_016864411.1 | 12 | 1814 | |||
XM_017008923.1 | XP_016864412.1 | 12 | 1783 | |||
GenBank mRNA | AK130003.1 | - | 9 | 1023 | ||
AK222606.1 | - | 12 | 1500 | |||
AK303580.1 | - | 12 | 1596 | |||
AK312572.1 | - | 12 | 1502 | |||
BC006997.1 | - | 12 | 1502 | 3950738 | ||
BC108278.1 | - | 12 | 1447 | 6051310 | ||
BT019384.1 | - | 11 | 1368 | |||
BX647869.1 | - | 11 | 1410 | |||
BX648012.1 | - | 12 | 1514 | |||
CR536535.1 | - | 11 | 1368 | |||
U32519.1 | - | 12 | 1489 |