Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KMT2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197104.1 | NP_001184033.1 | 3 | 748 | ||
NM_005933.3 | NP_005924.2 | 3 | 748 | |||
XM_011542829.2 | XP_011541131.1 | 4 | 839 | |||
XM_011542830.2 | XP_011541132.1 | 4 | 839 | |||
XM_011542831.2 | XP_011541133.1 | 4 | 839 | |||
GenBank mRNA | D14540.1 | - | 3 | 748 | ||
L04284.1 | - | 3 | 725 | |||
L04731.1 | - | 3 | 539 |