Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIRT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193286.1 | NP_001180215.1 | 9, 10 | 989 | ||
NM_012237.3 | NP_036369.2 | 10, 11 | 997 | |||
NM_030593.2 | NP_085096.1 | 9, 10 | 989 | |||
NR_034146.1 | 8, 9 | 920 | ||||
XM_006723111.1 | XP_006723174.1 | 11, 12 | 945 | |||
XM_011526654.1 | XP_011524956.1 | 9, 10 | 882 | |||
XM_011526655.1 | XP_011524957.1 | 7, 8 | 697 | |||
XM_017026500.1 | XP_016881989.1 | 9, 10 | 921 | |||
GenBank mRNA | AB009274.1 | - | 9, 10 | 710 | ||
AF083107.2 | - | 10, 11 | 895 | |||
AF095714.1 | - | 9, 10 | 768 | |||
AF131800.1 | - | 8, 9 | 644 | |||
AF160214.1 | - | 9, 10 | 749 | |||
AJ505014.1 | - | 7, 8 | 1466 | |||
AK025876.1 | - | 9, 10 | 775 | |||
AK054642.1 | - | 10, 11 | 828 | |||
AK225824.1 | - | 9, 10 | 772 | |||
AK290716.1 | - | 8, 9 | 920 | |||
AK314492.1 | - | 10, 11 | 751 | |||
AY030277.1 | - | 9, 10 | 750 | |||
BC003012.1 | - | 9, 10 | 840 | 2820929 | ||
BC003547.1 | - | 9, 10 | 840 | 3536728 | ||
KF032391.1 | - | 7, 8 | 697 |