Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320687.1 | NP_001307616.1 | 3 | 1014 | ||
NM_016166.2 | NP_057250.1 | 2 | 415 | |||
XM_011522126.2 | XP_011520428.1 | 4 | 1122 | |||
XM_011522127.2 | XP_011520429.1 | 2 | 899 | |||
XM_017022688.1 | XP_016878177.1 | 2 | 893 | |||
XM_017022689.1 | XP_016878178.1 | 2 | 906 | |||
XM_017022690.1 | XP_016878179.1 | 2 | 711 | |||
XM_017022691.1 | XP_016878180.1 | 2 | 485 | |||
GenBank mRNA | AF077951.1 | - | 2 | 267 | ||
AF167160.1 | - | 2 | 363 | |||
AK094641.1 | - | 3 | 1014 | |||
AK314515.1 | - | 2 | 290 | |||
AY826819.1 | - | 2 | 264 | |||
BC053685.1 | - | 2 | 268 | |||
BC104646.1 | - | 2 | 283 | |||
BC118587.1 | - | 2 | 317 | 40085010 | ||
BC121797.1 | - | 2 | 314 | 40085007 | ||
EF487997.1 | - | 2 | 267 | |||
EF487998.1 | - | 2 | 267 | |||
FJ997900.1 | - | 2 | 267 | |||
U78524.1 | - | 1 | 249 |