Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDAC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166418.1 | NP_001159890.1 | 9 | 1199 | ||
NM_018486.2 | NP_060956.1 | 11 | 1472 | |||
NR_051952.1 | 10 | 1330 | ||||
XM_011530986.2 | XP_011529288.2 | 12 | 1545 | |||
XM_017029640.1 | XP_016885129.1 | 11 | 1467 | |||
XM_017029641.1 | XP_016885130.1 | 10 | 1389 | |||
XM_017029642.1 | XP_016885131.1 | 10 | 1270 | |||
XM_017029643.1 | XP_016885132.1 | 9 | 1245 | |||
XM_017029644.1 | XP_016885133.1 | 9 | 1194 | |||
XM_017029645.1 | XP_016885134.1 | 8 | 1167 | |||
XM_017029646.1 | XP_016885135.1 | 11 | 1094 | |||
GenBank mRNA | AF230097.1 | - | 11 | 1185 | ||
AF245664.1 | - | 11 | 1139 | |||
AJ277724.1 | - | 11 | 1172 | |||
AK296641.1 | - | 9 | 1149 | |||
AK298767.1 | - | 10 | 1070 | |||
BC050433.1 | - | 11 | 1163 | 5761745 |