Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KMT5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324504.1 | NP_001311433.1 | 9 | 1363 | ||
NM_001324505.1 | NP_001311434.1 | 7 | 1331 | |||
NM_001324506.1 | NP_001311435.1 | 6 | 1237 | |||
NM_020382.4 | NP_065115.3 | 8 | 1453 | |||
NR_136910.1 | 9 | 1566 | ||||
XM_006719394.3 | XP_006719457.1 | 7 | 1283 | |||
XM_011538333.2 | XP_011536635.1 | 7 | 1669 | |||
GenBank mRNA | AK292645.1 | - | 8 | 1421 | ||
AY064546.1 | - | 1 | 1506 | |||
AY102937.1 | - | 8 | 1383 | |||
BC050346.1 | - | 8 | 1794 | 4838030 |