Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMYD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167740.1 | NP_001161212.1 | 10, 11 | 1201 | ||
NM_022743.2 | NP_073580.1 | 10, 11 | 1029 | |||
XM_011544253.1 | XP_011542555.1 | 9, 10 | 1066 | |||
XM_011544254.2 | XP_011542556.1 | 6, 7 | 1173 | |||
XM_011544257.1 | XP_011542559.1 | 6, 7 | 665 | |||
XM_011544258.2 | XP_011542560.1 | 7, 8 | 1022 | |||
XM_011544259.2 | XP_011542561.1 | 6, 7 | 888 | |||
XM_011544260.1 | XP_011542562.1 | 6, 7 | 705 | |||
XM_011544261.2 | XP_011542563.1 | 6, 7 | 630 | |||
XM_017002094.1 | XP_016857583.1 | 6, 7 | 4845 | |||
XM_017002095.1 | XP_016857584.1 | 6, 7 | 576 | |||
GenBank mRNA | AB057595.1 | - | 10, 11 | 1171 | ||
AK023594.1 | - | 5, 6 | 1431 | |||
AK024733.1 | - | 10, 11 | 1026 | |||
AK289605.1 | - | 10, 11 | 1177 | |||
AY186742.1 | - | 7, 8 | 940 | |||
BC017079.1 | - | 6, 7 | 653 | |||
BC031010.1 | - | 10, 11 | 1116 | 4334047 |