Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SETD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030648.3 | NP_085151.1 | 8 | 2305 | ||
XM_017008661.1 | XP_016864150.1 | 6 | 1398 | |||
GenBank mRNA | AB051504.1 | - | 8 | 1940 | ||
AF448510.1 | - | 8 | 1940 | |||
BC121055.1 | - | 8 | 1849 | 40118363 | ||
BC121056.1 | - | 8 | 1849 | 40118365 |