Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITCH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257137.2 | NP_001244066.1 | 26 | 4498 | ||
NM_001257138.2 | NP_001244067.1 | 24 | 4233 | |||
NM_001324197.1 | NP_001311126.1 | 26 | 4519 | |||
NM_001324198.1 | NP_001311127.1 | 25 | 4396 | |||
NM_031483.6 | NP_113671.3 | 25 | 4375 | |||
XM_005260570.1 | XP_005260627.1 | 24 | 4291 | |||
XM_011529079.1 | XP_011527381.1 | 24 | 4311 | |||
XM_017028089.1 | XP_016883578.1 | 25 | 4331 | |||
XM_017028090.1 | XP_016883579.1 | 26 | 4451 | |||
XM_017028091.1 | XP_016883580.1 | 24 | 4246 |