Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AURKA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323303.1 | NP_001310232.1 | 9 | 1459 | ||
NM_001323304.1 | NP_001310233.1 | 9 | 1222 | |||
NM_001323305.1 | NP_001310234.1 | 10 | 1332 | |||
NM_003600.3 | NP_003591.2 | 8 | 1349 | |||
NM_198433.2 | NP_940835.1 | 10 | 1557 | |||
NM_198434.2 | NP_940836.1 | 9 | 1248 | |||
NM_198435.2 | NP_940837.1 | 8 | 1138 | |||
NM_198436.2 | NP_940838.1 | 9 | 1234 | |||
NM_198437.2 | NP_940839.1 | 8 | 1124 | |||
XM_017028034.1 | XP_016883523.1 | 10 | 1679 | |||
XM_017028035.1 | XP_016883524.1 | 8 | 1039 | |||
GenBank mRNA | AF008551.1 | - | 7 | 949 | ||
AF011468.1 | - | 8 | 1205 | |||
AK301769.1 | - | 8 | 811 | |||
AK312776.1 | - | 8 | 1016 | |||
BC001280.1 | - | 9 | 1081 | 3456017 | ||
BC002499.2 | - | 8 | 1077 | 3051177 | ||
BC006423.1 | - | 8 | 974 | 3532438 | ||
BC027464.1 | - | 9 | 1116 | 4449474 | ||
D84212.1 | - | 9 | 1115 |