Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AURKA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323303.1 | NP_001310232.1 | 9 | 1526 | ||
NM_001323304.1 | NP_001310233.1 | 9 | 1289 | |||
NM_001323305.1 | NP_001310234.1 | 10 | 1399 | |||
NM_003600.3 | NP_003591.2 | 8 | 1416 | |||
NM_198433.2 | NP_940835.1 | 10 | 1624 | |||
NM_198434.2 | NP_940836.1 | 9 | 1315 | |||
NM_198435.2 | NP_940837.1 | 8 | 1205 | |||
NM_198436.2 | NP_940838.1 | 9 | 1301 | |||
NM_198437.2 | NP_940839.1 | 8 | 1191 | |||
XM_017028034.1 | XP_016883523.1 | 10 | 1746 | |||
XM_017028035.1 | XP_016883524.1 | 8 | 1106 | |||
GenBank mRNA | AF008551.1 | - | 7 | 1016 | ||
AF011468.1 | - | 8 | 1272 | |||
AK301769.1 | - | 8 | 878 | |||
AK312776.1 | - | 8 | 1083 | |||
BC001280.1 | - | 9 | 1148 | 3456017 | ||
BC002499.2 | - | 8 | 1144 | 3051177 | ||
BC006423.1 | - | 8 | 1041 | 3532438 | ||
BC027464.1 | - | 9 | 1183 | 4449474 | ||
D84212.1 | - | 9 | 1182 |
Gene Symbol: AURKAPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001587.1 | 1 | 814 |