Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2J1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016021.2 | NP_057105.2 | 8 | 1750 | ||
XM_011535887.1 | XP_011534189.1 | 7 | 1617 | |||
XM_011535888.2 | XP_011534190.1 | 8 | 1797 | |||
GenBank mRNA | AF151834.1 | - | 8 | 1557 | ||
AJ420578.1 | - | 7 | 1399 |