Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WHSC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042424.2 | NP_001035889.1 | 2 | 650 | ||
NM_007331.1 | NP_015627.1 | 4 | 968 | |||
NM_133330.2 | NP_579877.1 | 4 | 968 | |||
NM_133331.2 | NP_579878.1 | 3 | 788 | |||
NM_133334.2 | NP_579889.1 | 1 | 503 | |||
NM_133335.3 | NP_579890.1 | 2 | 613 | |||
XM_005248001.3 | XP_005248058.1 | 2 | 593 | |||
XM_005248002.2 | XP_005248059.1 | 2 | 689 | |||
XM_005248005.2 | XP_005248062.1 | 2 | 689 | |||
XM_006713914.3 | XP_006713977.1 | 3 | 858 | |||
XM_011513557.2 | XP_011511859.1 | 2 | 529 | |||
GenBank mRNA | AF071593.1 | - | 1 | 503 | ||
AF071594.1 | - | 1 | 503 | |||
AF083386.1 | - | 4 | 968 | |||
AF083387.1 | - | 3 | 788 | |||
AF083388.1 | - | 2 | 612 | |||
AF083389.1 | - | 4 | 968 | |||
AF083390.1 | - | 4 | 968 | |||
AF083391.1 | - | 4 | 968 | |||
AJ007042.1 | - | 2 | 1236 | |||
AK289697.1 | - | 3 | 635 | |||
AY694128.1 | - | 1 | 485 | |||
BC020545.1 | - | 2 | 585 | |||
BC052254.1 | - | 2 | 592 | |||
BC141815.1 | - | 2 | 683 | |||
CX866583.1 | - | 2 | 679 |