Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHEK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114121.2 | NP_001107593.1 | 13 | 2288 | ||
NM_001114122.2 | NP_001107594.1 | 13 | 2288 | |||
NM_001244846.1 | NP_001231775.1 | 12 | 2186 | |||
NM_001274.5 | NP_001265.2 | 13 | 1631 | |||
NR_045204.1 | 12 | 2064 | ||||
NR_045205.1 | 13 | 1825 | ||||
XM_011542560.2 | XP_011540862.1 | 12 | 2290 | |||
XM_011542561.2 | XP_011540863.1 | 12 | 2290 | |||
XM_011542562.2 | XP_011540864.1 | 12 | 1540 | |||
XM_011542563.2 | XP_011540865.1 | 12 | 1605 | |||
XM_017017146.1 | XP_016872635.1 | 13 | 1475 | |||
GenBank mRNA | AB451222.1 | - | 12 | 1391 | ||
AB451345.1 | - | 12 | 1391 | |||
AF016582.1 | - | 12 | 1425 | |||
AF032874.1 | - | 12 | 1391 | |||
AK292549.1 | - | 13 | 1825 | |||
AK293143.1 | - | 12 | 1351 | |||
AK299783.1 | - | 12 | 1225 | |||
AK300082.1 | - | 12 | 1474 | |||
BC004202.2 | - | 13 | 1450 | 3530606 | ||
BC017575.1 | - | 13 | 1549 | 4091997 | ||
BG191840.1 | - | 1 | 58 | |||
BG216281.1 | - | 1 | 55 | |||
BG218896.1 | - | 2 | 162 | |||
JF289264.1 | - | 12 | 1266 |