Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202476.1 | NP_001189405.1 | 5 | 505 | ||
NM_003341.4 | NP_003332.1 | 6 | 800 | |||
NM_182666.2 | NP_872607.1 | 5 | 749 | |||
GenBank mRNA | AK314854.1 | - | 6 | 709 | ||
BC009139.1 | - | 6 | 724 | 3853408 | ||
BI223271.1 | - | 5 | 644 | |||
X92963.1 | - | 5 | 571 |