Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDAC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003883.3 | NP_003874.2 | 15 | 1818 | ||
XM_011537697.2 | XP_011535999.1 | 14 | 1670 | |||
GenBank mRNA | AF005482.1 | - | 13 | 1835 | ||
AF039703.1 | - | 15 | 1774 | |||
BC000614.1 | - | 15 | 1807 | 3345755 | ||
U66914.1 | - | 15 | 1764 | |||
U75696.1 | - | 14 | 1802 | |||
U75697.1 | - | 15 | 1808 |