Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMYD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167740.1 | NP_001161212.1 | 3 | 379 | ||
NM_022743.2 | NP_073580.1 | 3 | 207 | |||
XM_011544253.1 | XP_011542555.1 | 3 | 355 | |||
GenBank mRNA | AB057595.1 | - | 3 | 349 | ||
AK024733.1 | - | 3 | 204 | |||
AK289605.1 | - | 3 | 355 | |||
BC031010.1 | - | 3 | 294 | 4334047 | ||
BC107725.1 | - | 3 | 353 | |||
BI916760.1 | - | 3 | 271 | |||
BP231249.1 | - | 3 | 378 |